Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004958.3 | NP_004949.1 | 12 | 2106 | |
XM_005263438.2 | XP_005263495.1 | 12 | 2113 | ||
XM_011541166.2 | XP_011539468.1 | 12 | 2106 | ||
XM_017000900.1 | XP_016856389.1 | 9 | 1569 | ||
XM_017000901.1 | XP_016856390.1 | 10 | 1857 | ||
XM_017000902.1 | XP_016856391.1 | 12 | 2106 | ||
GenBank mRNA | AB209995.1 | - | 12 | 2087 | |
AK302863.1 | - | 12 | 2098 | ||
AK304273.1 | - | 12 | 2095 | ||
BC117166.1 | - | 11 | 2003 | 40125717 | |
L34075.1 | - | 12 | 2064 | ||
U88966.1 | - | 12 | 2064 |