Gene Transcripts

Gene Symbol
GPR25
Entrez Gene ID
2848
Gene Name
G protein-coupled receptor 25
Gene Aliases
Chromosome Location
Chr. 1: 200872955 - 200874178 on Build GRCh38
UniGene ID
Hs.534316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005298.3 NP_005289.2 1 1118
GenBank mRNA AK315339.1 - 1 1092
BC126386.1 - 1 1118 8992102
BC126388.1 - 1 1118 8992104

View Full Product Details