Gene Transcripts

Gene Symbol
Abi1
Entrez Gene ID
11308
Gene Name
abl-interactor 1
Gene Aliases
E3B1, NAP1, Ssh3bp1
Chromosome Location
Chr. 2: 22939989 - 23040241 on Build GRCm38
UniGene ID
Mm.205647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077190.2 NP_001070658.1 3 588
NM_001077192.2 NP_001070660.1 3 588
NM_001077193.2 NP_001070661.1 3 588
NM_007380.3 NP_031406.2 3 588
NM_145994.2 NP_666106.1 3 588
GenBank mRNA AF420251.1 - 3 524
AK034476.1 - 3 470
AK151026.1 - 3 496
AK151363.1 - 3 567
AK152046.1 - 1 508
AK152061.1 - 3 452
AK152184.1 - 3 452
AK153130.1 - 3 496
AK164040.1 - 3 504
AK165924.1 - 3 561
AK167328.1 - 3 547
AK167568.1 - 3 543
AK167744.1 - 3 535
AK181529.1 - 1 116
AK184875.1 - 1 32
AK190191.1 - 1 32
AK199630.1 - 1 32
AK201770.1 - 1 32
AK215001.1 - 1 32
AY033645.1 - 3 432
BC004657.1 - 3 539 3498068
BC079642.1 - 3 494 6816252
U17698.1 - 2 214

View Full Product Details