Gene Transcripts

Gene Symbol
Aco2
Entrez Gene ID
11429
Gene Name
aconitase 2, mitochondrial
Gene Aliases
Aco-2, Aco3, D10Wsu183e
Chromosome Location
Chr. 15: 81872463 - 81915137 on Build GRCm38
UniGene ID
Mm.154581
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080633.2 NP_542364.1 12 1479
GenBank mRNA AK143917.1 - 12 1458
AK144207.1 - 12 1442
AK145511.1 - 12 1459
AK150027.1 - 12 1459
AK150954.1 - 13 1441
AK165411.1 - 12 1443
BC004645.1 - 12 1505 3158239
BC094462.1 - 12 1438 4238599
CO426953.1 - 5 413

View Full Product Details