Gene Transcripts

Gene Symbol
Cd3e
Entrez Gene ID
12501
Gene Name
CD3 antigen, epsilon polypeptide
Gene Aliases
AI504783, CD3, CD3epsilon, T3e
Chromosome Location
Chr. 9: 44998735 - 45009662 on Build GRCm38
UniGene ID
Mm.210361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007648.4 NP_031674.1 5 352
GenBank mRNA BC145926.1 - 5 352
BC145954.1 - 5 352 40131257
GU456631.1 - 4 274
J02990.1 - 5 353

View Full Product Details