Gene Transcripts

Gene Symbol
Cmas
Entrez Gene ID
12764
Gene Name
cytidine monophospho-N-acetylneuraminic acid synthetase
Gene Aliases
AW208911, CMPNeu5Ac, D6Bwg0250e
Chromosome Location
Chr. 6: 142756686 - 142775714 on Build GRCm38
UniGene ID
Mm.3820
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009908.2 NP_034038.2 2, 3 479
GenBank mRNA AJ006215.1 - 2, 3 392
AK155730.1 - 2, 3 430
BC031500.1 - 2, 3 408 2647633
BC063776.1 - 2, 3 408 6398450

View Full Product Details