Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016706.2 | NP_057915.2 | 3 | 1396 | |
GenBank mRNA | AF208663.1 | - | 3 | 1378 | |
AK146366.1 | - | 3 | 1380 | ||
BC031167.1 | - | 3 | 1411 | 4983919 | |
CF105200.1 | - | 2 | 194 |