Gene Transcripts
- Gene Symbol
- H3F3A
- Entrez Gene ID
- 3020
- Gene Name
- H3 histone, family 3A
- Gene Aliases
- H3.3A, H3F3
- Chromosome Location
- Chr. 1: 226062707 - 226072002 on Build GRCh38
- UniGene ID
- Hs.533624
- Species
- Homo sapiens
- Gene Symbol
- H3F3AP2
- Entrez Gene ID
- 664611
- Gene Name
- H3 histone, family 3A pseudogene 2
- Gene Aliases
- H3F3CP
- Chromosome Location
- Chr. 14: 23294524 - 23295800 on Build GRCh38
- UniGene ID
- Hs.710438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057381.1 |
- |
1 |
1456 |
|
- Gene Symbol
- H3F3AP4
- Entrez Gene ID
- 440926
- Gene Name
- H3 histone, family 3A, pseudogene 4
- Gene Aliases
- p13
- Chromosome Location
- Chr. 2: 174719800 - 174720858 on Build GRCh38
- UniGene ID
- Hs.533624
- Species
- Homo sapiens
View Full Product Details