Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037811.2 | NP_001032900.1 | 4 | 413 | |
NM_004493.2 | NP_004484.1 | 4 | 413 | ||
GenBank mRNA | AF035555.2 | - | 4 | 418 | |
AF069134.1 | - | 4 | 400 | ||
AU132847.1 | - | 4 | 413 | ||
AY092415.1 | - | 3 | 232 | ||
BC000372.2 | - | 4 | 397 | 2819721 | |
BC000829.1 | - | 4 | 397 | ||
BC008708.1 | - | 4 | 393 | 3160844 | |
CR456723.1 | - | 4 | 382 | ||
KU178107.1 | - | 4 | 382 | ||
KU178108.1 | - | 4 | 382 | ||
U73514.1 | - | 4 | 397 | ||
U96132.1 | - | 4 | 409 |