Gene Transcripts
- Gene Symbol
- Crygf
- Entrez Gene ID
- 12969
- Gene Name
- crystallin, gamma F
- Gene Aliases
- 3110001K11Rik, Cryg-2, DGcry-2, Len-2
- Chromosome Location
- Chr. 1: 65926522 - 65928304 on Build GRCm38
- UniGene ID
- Mm.275362
- Species
- Mus musculus
- Gene Symbol
- Cryge
- Entrez Gene ID
- 12968
- Gene Name
- crystallin, gamma E
- Gene Aliases
- Cryg-2, Cryg-6, DGcry-2, Elo
- Chromosome Location
- Chr. 1: 65048555 - 65057116 on Build GRCm38
- UniGene ID
- Mm.258170
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
K02584.1 |
- |
|
284 |
|
View Full Product Details