Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010016.2 | NP_034146.2 | 8 | 1104 | |
GenBank mRNA | AK030285.1 | - | 8 | 2007 | |
AK160994.1 | - | 8 | 1082 | ||
BC011314.1 | - | 8 | 1059 | 3663561 | |
CT010328.1 | - | 8 | 1033 | ||
D63679.1 | - | 8 | 1015 | ||
L41366.1 | - | 8 | 1048 |