Gene Transcripts

Gene Symbol
Emp3
Entrez Gene ID
13732
Gene Name
epithelial membrane protein 3
Gene Aliases
H-4, H4, HNMP-1, MI-35, Ymp
Chromosome Location
Chr. 7: 45918023 - 45921426 on Build GRCm38
UniGene ID
Mm.20829
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146346.1 NP_001139818.1 3 384
NM_010129.2 NP_034259.2 3 355
GenBank mRNA AF011750.1 - 3 216
AK146939.1 - 3 387
BC001999.1 - 3 200 3485948
CK022835.1 - 3 354
U87948.1 - 3 333

View Full Product Details