Gene Transcripts

Gene Symbol
Ptprv
Entrez Gene ID
13924
Gene Name
protein tyrosine phosphatase, receptor type, V
Gene Aliases
Esp, OST, OST-PTP, mOST-PTP
Chromosome Location
Chr. 1: 135108498 - 135132575 on Build GRCm38
UniGene ID
Mm.4450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007955.3 NP_031981.3 10 1962
GenBank mRNA AF300701.1 - 9 1439
AK156112.1 - 10 1964
U36488.1 - 6 1124

View Full Product Details