Gene Transcripts

Gene Symbol
Ezh2
Entrez Gene ID
14056
Gene Name
enhancer of zeste homolog 2 (Drosophila)
Gene Aliases
Enx-1, Enx1h, KMT6, mKIAA4065
Chromosome Location
Chr. 6: 47530274 - 47595340 on Build GRCm38
UniGene ID
Mm.246688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146689.1 NP_001140161.1 12 1553
NM_007971.2 NP_031997.2 12 1565
GenBank mRNA AK157853.1 - 12 1566
AK220174.1 - 12 1520
BC003772.1 - 12 1474 3492110
BC016391.1 - 12 1487 3586689
BC079538.1 - 12 1504 6817366
U52951.1 - 12 1555

View Full Product Details