Gene Transcripts

Gene Symbol
Fgfr1
Entrez Gene ID
14182
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
AW208770, Eask, FLG, Fgfr-1, Flt-2, Hspy, MFR, bFGF-R-1, c-fgr
Chromosome Location
Chr. 8: 25513623 - 25575718 on Build GRCm38
UniGene ID
Mm.265716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079908.2 NP_001073377.1 14 2651
NM_001079909.2 NP_001073378.1 13 2390
NM_010206.3 NP_034336.2 14 2657
GenBank mRNA AF176552.1 - 12 1688
AK028354.1 - 13 1830
AK034295.1 - 4 477
AK049704.1 - 14 2617
AK167639.1 - 5 1954
BC010200.1 - 14 2550 3967874
BC033447.1 - 12 1653 4037899
BC053921.1 - 7 1897
M28998.1 - 13 1955
M33760.1 - 12 1674
M65053.1 - 13 1928
U22324.1 - 13 1955
U23445.1 - 12 1688
X51893.1 - 14 2012

View Full Product Details