Gene Transcripts

Gene Symbol
Ganab
Entrez Gene ID
14376
Gene Name
alpha glucosidase 2 alpha neutral subunit
Gene Aliases
AU042638, G2an, GluII, mKIAA0088
Chromosome Location
Chr. 19: 8898057 - 8916742 on Build GRCm38
UniGene ID
Mm.3196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293621.1 NP_001280550.1 9 1028
NM_008060.2 NP_032086.1 10 1094
GenBank mRNA AK028655.1 - 9 998
AK030722.1 - 9 997
AK047454.1 - 4 349
AK081915.1 - 9 990
AK122201.1 - 4 1224
AK149686.1 - 10 1051
AK166150.1 - 9 967
BC020386.1 - 2 244
BC094437.1 - 9 978 30682580
BC117888.1 - 9 971 40096413
BC117889.1 - 9 974 40096414
BC128069.1 - 10 1038 40096428
U92793.1 - 10 1040

Gene Symbol
Efna4
Entrez Gene ID
13639
Gene Name
ephrin A4
Gene Aliases
EFL-4, Epl4, LERK-4
Chromosome Location
Chr. 3: 89333393 - 89338028 on Build GRCm38
UniGene ID
Mm.16332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA U90663.1 - 38

View Full Product Details