Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008078.2 | NP_032104.2 | 11 | 1453 | |
GenBank mRNA | AF326549.1 | - | 11 | 1117 | |
AF326550.1 | - | 11 | 1117 | ||
AK136834.1 | - | 11 | 1153 | ||
BC018380.1 | - | 11 | 1168 | 4482097 | |
D42051.1 | - | 11 | 1469 | ||
KC134211.1 | - | 11 | 1117 | ||
L16980.1 | - | 11 | 1189 |