Gene Transcripts

Gene Symbol
Gata6
Entrez Gene ID
14465
Gene Name
GATA binding protein 6
Gene Aliases
AA410133, GATA-6
Chromosome Location
Chr. 18: 11052475 - 11085636 on Build GRCm38
UniGene ID
Mm.329287
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010258.3 NP_034388.2 4 1667
GenBank mRNA AF179425.1 - 3 974
AK142381.1 - 4 1527
S82462.1 - 3 1429
U51335.1 - 2 247

View Full Product Details