Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010294.1 | NP_034424.1 | 1 | 1491 | |
GenBank mRNA | AF117734.1 | - | 1 | 1491 | |
AK075761.1 | - | 1 | 1404 | ||
BC050764.1 | - | 1 | 1504 | 6774427 | |
BC061147.1 | - | 1 | 1470 | 6770827 |