Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290821.1 | NP_001277750.1 | 3, 4 | 714 | |
NM_020492.4 | NP_065238.2 | 3, 4 | 714 | ||
GenBank mRNA | AK078258.1 | - | 3, 4 | 714 | |
AY129229.1 | - | 3, 4 | 261 | ||
BC131949.1 | - | 3, 4 | 379 | 40130226 | |
BC137725.1 | - | 3, 4 | 379 | 8860744 | |
S73717.1 | - | 3, 4 | 306 | ||
S73718.1 | - | 3, 4 | 306 |