Gene Transcripts

Gene Symbol
Hnrnpa1
Entrez Gene ID
15382
Gene Name
heterogeneous nuclear ribonucleoprotein A1
Gene Aliases
HDP-1, Hdp, Hnrpa1, hnRNP A1, hnrnp-A1
Chromosome Location
Chr. 15: 103240397 - 103246698 on Build GRCm38
UniGene ID
Mm.237064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039129.4 NP_001034218.1 9 1105
NM_010447.5 NP_034577.1 8 946
GenBank mRNA AK007802.1 - 8 892
AK088308.1 - 8 891
AK131999.1 - 8 912
AK135391.1 - 8 890
AK136113.1 - 8 916
AK146126.1 - 9 1049
AK146630.1 - 9 1071
AK152684.1 - 8 890
AK161730.1 - 8 890
AK166901.1 - 8 890
AK166989.1 - 9 1047
AK167161.1 - 8 890
AK167810.1 - 8 890
AK167811.1 - 8 890
AK167820.1 - 8 891
AK167913.1 - 8 890
AK168420.1 - 8 890
AK168531.1 - 8 893
AK168733.1 - 9 1049
AK169216.1 - 8 890
BC052723.1 - 9 1035
BC080675.1 - 8 872 6308746
BC083136.1 - 8 857 6511966
BC089340.1 - 9 1024 6402927
BC092395.1 - 9 1025 6403376
D86729.1 - 8 853
M99167.1 - 8 888

View Full Product Details