Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001122950.2 | NP_001116422.1 | 2 | 700 | |
NM_008263.3 | NP_032289.2 | 2 | 1104 | ||
GenBank mRNA | AK002670.1 | - | 2 | 252 | |
AK188006.1 | - | 1 | 73 | ||
BC050839.1 | - | 2 | 1076 | 6511608 | |
L08757.1 | - | 2 | 1104 |