Gene Transcripts

Gene Symbol
HOXA6
Entrez Gene ID
3203
Gene Name
homeobox A6
Gene Aliases
HOX1, HOX1.2, HOX1B
Chromosome Location
Chr. 7: 27145583 - 27147774 on Build GRCh38
UniGene ID
Hs.679517
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024014.3 NP_076919.1 1 196
GenBank mRNA BC069497.1 - 1 171
BC104915.1 - 1 196 8143918
BC104917.1 - 1 196 8143920

View Full Product Details