Gene Transcripts

Gene Symbol
Hoxc6
Entrez Gene ID
15425
Gene Name
homeobox C6
Gene Aliases
Hox-3.3, Hox-6.1
Chromosome Location
Chr. 15: 102998226 - 103011882 on Build GRCm38
UniGene ID
Mm.4444
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010465.2 NP_034595.2 1 296
GenBank mRNA AB221685.1 - 1 252
AK050798.1 - 1 64
BC028482.1 - 1 296
BC141061.1 - 1 275 9055673
BX528590.1 - 1 297
X12504.1 - 2 321
X16510.1 - 257
X16511.1 - 1 379

View Full Product Details