Gene Transcripts

Gene Symbol
Hoxd10
Entrez Gene ID
15430
Gene Name
homeobox D10
Gene Aliases
AI385591, AI874987, Hox-4.5, Hox-5.3
Chromosome Location
Chr. 2: 74684275 - 74695106 on Build GRCm38
UniGene ID
Mm.24420
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013554.5 NP_038582.2 1 354
GenBank mRNA AK134330.1 - 1 355
BC013463.1 - 1 315 4240579
BC048690.1 - 1 297 6516538

View Full Product Details