Gene Transcripts
- Gene Symbol
- HOXA10
- Entrez Gene ID
- 3206
- Gene Name
- homeobox A10
- Gene Aliases
- HOX1, HOX1.8, HOX1H, PL
- Chromosome Location
- Chr. 7: 27170591 - 27180261 on Build GRCh38
- UniGene ID
- Hs.110637
- Species
- Homo sapiens
- Gene Symbol
- HOXA10-HOXA9
- Entrez Gene ID
- 100534589
- Gene Name
- HOXA10-HOXA9 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 7: 27162438 - 27180261 on Build GRCh38
- UniGene ID
- Hs.659350
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
FY213345.1 |
- |
2 |
174 |
|
View Full Product Details