Gene Transcripts

Gene Symbol
Irx3
Entrez Gene ID
16373
Gene Name
Iroquois related homeobox 3
Gene Aliases
AI894186
Chromosome Location
Chr. 8: 91798511 - 91801654 on Build GRCm38
UniGene ID
Mm.238044
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008393.3 NP_032419.2 2, 3 1808
GenBank mRNA BC067195.1 - 2, 3 1807
BC085500.1 - 2, 3 1528 30539191
Y15001.1 - 2, 3 1394

View Full Product Details