Gene Transcripts

Gene Symbol
Itga2b
Entrez Gene ID
16399
Gene Name
integrin alpha 2b
Gene Aliases
AI172977, CD41, CD41B, GpIIb, alphaIIb
Chromosome Location
Chr. 11: 102453297 - 102469883 on Build GRCm38
UniGene ID
Mm.26646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010575.2 NP_034705.2 13 1550
GenBank mRNA AF166384.1 - 13 1428
AF170316.1 - 13 1411
AK048700.1 - 8 1058
AK140749.1 - 8 1202
AK142289.1 - 13 1412
AK154619.1 - 13 1551
AK156872.1 - 12 1732
AK157012.1 - 12 1734
AK158443.1 - 13 1397
BC120493.1 - 13 1400 40129416
BC137570.1 - 13 1400 8860589

View Full Product Details