Gene Transcripts

Gene Symbol
Klk1b1
Entrez Gene ID
16623
Gene Name
kallikrein 1-related peptidase b1
Gene Aliases
Klk1, TK, mGK-1, mK1
Chromosome Location
Chr. 7: 43966768 - 43971316 on Build GRCm38
UniGene ID
Mm.447819
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010645.2 NP_034775.1 3 503
GenBank mRNA AK089370.1 - 3 521
BC026378.1 - 3 503 4165617

View Full Product Details