Gene Transcripts

Gene Symbol
Serpina3c
Entrez Gene ID
16625
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 3C
Gene Aliases
1A1, Kalbp, Klkbp, spi2
Chromosome Location
Chr. 12: 104146907 - 104153933 on Build GRCm38
UniGene ID
Mm.422814
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008458.2 NP_032484.1 2 193
GenBank mRNA BC116724.1 - 2 191

View Full Product Details