Gene Transcripts
- Gene Symbol
- Krtap12-1
- Entrez Gene ID
- 16694
- Gene Name
- keratin associated protein 12-1
- Gene Aliases
- D10Jhu14e
- Chromosome Location
- Chr. 10: 77720586 - 77721256 on Build GRCm38
- UniGene ID
- Mm.34372
- Species
- Mus musculus
- Gene Symbol
- Gm10024
- Entrez Gene ID
- 100009614
- Gene Name
- predicted gene 10024
- Gene Aliases
- ENSMUSG00000057573
- Chromosome Location
- Chr. 10: 77711446 - 77712009 on Build GRCm38
- UniGene ID
- Mm.35821
- Species
- Mus musculus
- Gene Symbol
- Gm10100
- Entrez Gene ID
- 100502953
- Gene Name
- predicted gene 10100
- Gene Aliases
- ENSMUSG00000061452
- Chromosome Location
- Chr. 10: 77726486 - 77726848 on Build GRCm38
- UniGene ID
- Mm.486157
- Species
- Mus musculus
- Gene Symbol
- Gm10142
- Entrez Gene ID
- 100502921
- Gene Name
- predicted gene 10142
- Gene Aliases
- ENSMUSG00000064028
- Chromosome Location
- Chr. 10: 77715807 - 77716169 on Build GRCm38
- UniGene ID
- Mm.486156
- Species
- Mus musculus
- Gene Symbol
- Gm10272
- Entrez Gene ID
- 16697
- Gene Name
- keratin associated protein 12-4
- Gene Aliases
- ENSMUSG00000069584, Krtap12-4
- Chromosome Location
- Chr. 10: 77706587 - 77706986 on Build GRCm38
- UniGene ID
- Mm.482251
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA079946.1 |
- |
1 |
81 |
|
View Full Product Details