Gene Transcripts

Gene Symbol
Lcp2
Entrez Gene ID
16822
Gene Name
lymphocyte cytosolic protein 2
Gene Aliases
AI323664, BB161688, SLP-76, SLP76, twm
Chromosome Location
Chr. 11: 34047201 - 34092280 on Build GRCm38
UniGene ID
Mm.265350
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010696.3 NP_034826.2 4 397
GenBank mRNA AK036727.1 - 4 455
AK037970.1 - 4 431
AK088552.1 - 4 519
AK153374.1 - 4 402
AK168134.1 - 4 413
AK170491.1 - 4 674
BC006948.1 - 4 397 3603181
U20159.1 - 4 436

View Full Product Details