Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161769.1 | NP_001155241.1 | 3 | 435 | |
NM_001161770.1 | NP_001155242.1 | 2 | 567 | ||
NM_010723.3 | NP_034853.1 | 3 | 722 | ||
GenBank mRNA | AF074600.1 | - | 2 | 283 | |
AF096996.1 | - | 3 | 451 | ||
AF102817.1 | - | 3 | 352 | ||
AK080534.1 | - | 3 | 370 | ||
AK138278.1 | - | 2 | 568 | ||
AK159590.1 | - | 3 | 649 | ||
BC003488.1 | - | 2 | 434 | 2811636 | |
BC004661.1 | - | 3 | 375 | ||
BC010278.1 | - | 3 | 465 | 3589532 | |
BC100382.1 | - | 3 | 327 | 6487478 |