Gene Transcripts

Gene Symbol
Klrb1a
Entrez Gene ID
17057
Gene Name
killer cell lectin-like receptor subfamily B member 1A
Gene Aliases
Ly55a, NKR-P1.7, NKR-P1A, NKRP12, Nkrp1-a, Nkrp1a, ly-55A
Chromosome Location
Chr. 6: 128609227 - 128622985 on Build GRCm38
UniGene ID
Mm.16729
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159902.1 NP_001153374.1 2 242
NM_010737.3 NP_034867.3 3 341
GenBank mRNA AF354261.1 - 3 240
AF354262.1 - 3 240
BC120707.1 - 3 279 40129630
BC145045.1 - 2 180 9053593
DQ237927.1 - 3 227
HQ713432.1 - 3 222
M77676.1 - 3 341
M77753.1 - 3 240
X64723.1 - 3 198

View Full Product Details