Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012235.3 | NP_036367.2 | 5 | 818 | |
XM_005264967.1 | XP_005265024.1 | 5 | 745 | ||
XM_011533501.1 | XP_011531803.1 | 5 | 845 | ||
XM_017005918.1 | XP_016861407.1 | 5 | 832 | ||
XM_017005919.1 | XP_016861408.1 | 5 | 765 | ||
XM_017005920.1 | XP_016861409.1 | 5 | 765 | ||
GenBank mRNA | BC027207.2 | - | 1 | 17 | |
D83782.2 | - | 5 | 804 |