Gene Transcripts

Gene Symbol
Mbd2
Entrez Gene ID
17191
Gene Name
methyl-CpG binding domain protein 2
Gene Aliases
MBD2a
Chromosome Location
Chr. 18: 70568270 - 70626131 on Build GRCm38
UniGene ID
Mm.322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010773.2 NP_034903.2 2 693
GenBank mRNA AF072243.1 - 2 651
AF072245.1 - 2 651
AK082825.1 - 2 129
AK147008.1 - 2 578
BC026698.1 - 2 653
BC046607.1 - 2 693 6399206
BC057289.1 - 2 525
BC058864.1 - 2 513

View Full Product Details