Gene Transcripts

Gene Symbol
Foxk1
Entrez Gene ID
17425
Gene Name
forkhead box K1
Gene Aliases
A630048H08Rik, AI463295, ENSMUSG00000075577, Gm10868, Mnf
Chromosome Location
Chr. 5: 142401497 - 142462015 on Build GRCm38
UniGene ID
Mm.24214
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199068.2 NP_951031.2 6 1377
GenBank mRNA AK144418.1 - 6 1270
AK147375.1 - 6 1375
BC060238.1 - 6 1357
L26507.1 - 6 1353

View Full Product Details