Gene Transcripts
- Gene Symbol
- GGT2
- Entrez Gene ID
- 728441
- Gene Name
- gamma-glutamyltransferase 2
- Gene Aliases
- GGT, GGT 2
- Chromosome Location
- Chr. 22: 21207972 - 21282982 on Build GRCh38
- UniGene ID
- Hs.568255
- Species
- Homo sapiens
- Gene Symbol
- IGSF3
- Entrez Gene ID
- 3321
- Gene Name
- immunoglobulin superfamily member 3
- Gene Aliases
- EWI-3, LCDD, V8
- Chromosome Location
- Chr. 1: 116574398 - 116667755 on Build GRCh38
- UniGene ID
- Hs.171057
- Species
- Homo sapiens
- Gene Symbol
- LOC102724197
- Entrez Gene ID
- 102724197
- Gene Name
- inactive gamma-glutamyltranspeptidase 2-like
- Gene Aliases
- GGT2
- Chromosome Location
- Chr. 22|NT_187386: 65299 - 86956 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details