Gene Transcripts

Gene Symbol
Nov
Entrez Gene ID
18133
Gene Name
nephroblastoma overexpressed gene
Gene Aliases
C130088N23Rik, CCN3
Chromosome Location
Chr. 15: 54745613 - 54753767 on Build GRCm38
UniGene ID
Mm.5167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010930.4 NP_035060.1 4 651
GenBank mRNA AK039481.1 - 4 642
AK081310.1 - 4 669
AK081944.1 - 4 877
AK136407.1 - 4 671
AK155048.1 - 4 670
BC003774.1 - 4 657 3494896
X96585.1 - 4 651
Y09257.1 - 4 639

View Full Product Details