Gene Transcripts

Gene Symbol
Prl2c4
Entrez Gene ID
26421
Gene Name
prolactin family 2, subfamily c, member 4
Gene Aliases
MRP-3, Mrpplf3, PLF-3, mrp/plf3
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.457984
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC064772.1 - 237

Gene Symbol
Prl2c3
Entrez Gene ID
18812
Gene Name
prolactin family 2, subfamily c, member 3
Gene Aliases
MRP-2, MRP-3, Mrpplf3, PLF-2, PLF-3, Plf2, Prl2c4, mrp/plf3
Chromosome Location
Chr. 13: 12790816 - 12800093 on Build GRCm38
UniGene ID
Mm.457984
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC132078.1 - 2 238
BC132080.1 - 2 238 40130357
K03235.1 - 2 255

View Full Product Details