Gene Transcripts

Gene Symbol
Pou2f2
Entrez Gene ID
18987
Gene Name
POU domain, class 2, transcription factor 2
Gene Aliases
Oct-2, Oct2a, Oct2b, Otf-2, Otf2
Chromosome Location
Chr. 7: 25091114 - 25179992 on Build GRCm38
UniGene ID
Mm.76683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163554.1 NP_001157026.1 12, 13 1165
NM_001163555.1 NP_001157027.1 12, 13 1231
NM_001163556.1 NP_001157028.1 12, 13 1165
NM_011138.2 NP_035268.2 12, 13 1213
GenBank mRNA AK156322.1 - 12, 13 1178
AY746973.1 - 12, 13 1193
AY746974.1 - 12, 13 1211
BC050258.1 - 12, 13 1178
BC104488.1 - 12, 13 1165 40046281
BC105647.1 - 12, 13 1165 40046279
BC105920.1 - 12, 13 1231 40046289
BC105921.1 - 12, 13 1165 40046286
S55236.1 - 5, 6 670
X53654.1 - 1373
X57089.1 - 5, 6 670
X57936.1 - 12, 13 1165
X57937.1 - 12, 13 1213
X57938.1 - 12, 13 1231
X57940.1 - 12, 13 1165
X57941.1 - 11, 12 1048

View Full Product Details