Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005534.3 | NP_005525.2 | 5 | 1346 | |
XM_005260969.2 | XP_005261026.1 | 6 | 862 | ||
XM_011529553.1 | XP_011527855.1 | 6 | 788 | ||
XM_011529554.2 | XP_011527856.1 | 5 | 1222 | ||
GenBank mRNA | AK292246.1 | - | 5 | 794 | |
AK314005.1 | - | 5 | 807 | ||
BC003624.2 | - | 5 | 814 | 2967074 | |
U05875.1 | - | 5 | 1346 | ||
U05877.1 | - | 5 | 811 |