Gene Transcripts

Gene Symbol
Psmb4
Entrez Gene ID
19172
Gene Name
proteasome (prosome, macropain) subunit, beta type 4
Gene Aliases
Pros-27
Chromosome Location
Chr. 3: 94884324 - 94886958 on Build GRCm38
UniGene ID
Mm.368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008945.3 NP_032971.2 2 365
GenBank mRNA AK077448.1 - 2 342
AK137044.1 - 2 341
AK156624.1 - 2 341
AK161348.1 - 2 341
AK168265.1 - 2 341
AK168340.1 - 2 341
BC008241.1 - 2 320 2654421
CB058553.1 - 2 365
M74556.1 - 1 143
U65636.1 - 2 320

View Full Product Details