Gene Transcripts
- Gene Symbol
- Rab1
- Entrez Gene ID
- 19324
- Gene Name
- RAB1, member RAS oncogene family
- Gene Aliases
- Gtbp, Rab-1, Rab1A, Ypt1, mKIAA3012
- Chromosome Location
- Chr. 11: 20201602 - 20226856 on Build GRCm38
- UniGene ID
- Mm.271944
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008996.3 |
NP_033022.1
|
2, 3 |
133 |
|
GenBank mRNA |
AB232583.1 |
- |
2, 3 |
105 |
|
AF226873.1 |
- |
2, 3 |
133 |
|
AK034408.1 |
- |
2, 3 |
301 |
|
AK050526.1 |
- |
2, 3 |
297 |
|
AK129477.1 |
- |
2, 3 |
291 |
|
AK151085.1 |
- |
2, 3 |
297 |
|
AK152978.1 |
- |
2, 3 |
302 |
|
AK159425.1 |
- |
2, 3 |
297 |
|
AK166249.1 |
- |
2, 3 |
294 |
|
AK167150.1 |
- |
2, 3 |
301 |
|
BC002077.1 |
- |
2, 3 |
300 |
3592802 |
Y00094.1 |
- |
2, 3 |
234 |
|
View Full Product Details