Gene Transcripts

Gene Symbol
Rasal1
Entrez Gene ID
19415
Gene Name
RAS protein activator like 1 (GAP1 like)
Gene Aliases
MRASAL
Chromosome Location
Chr. 5: 120648040 - 120679610 on Build GRCm38
UniGene ID
Mm.41209
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281999.1 NP_001268928.1 16 1973
NM_013832.4 NP_038860.2 17 1949
GenBank mRNA AF086714.1 - 16 1821
AK156050.1 - 16 1937
BC005418.1 - 17 1963 3488024

View Full Product Details