Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011246.2 | NP_035376.1 | 7 | 872 | |
GenBank mRNA | AF106070.1 | - | 7 | 716 | |
AK047613.1 | - | 2 | 174 | ||
AK134005.1 | - | 7 | 886 | ||
AK141524.1 | - | 7 | 997 | ||
BC057120.1 | - | 7 | 872 | 6831137 | |
BC057341.1 | - | 7 | 872 | 6831137 |