Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001139516.1 | NP_001132988.1 | 9 | 1283 | |
NM_011249.2 | NP_035379.2 | 9 | 1283 | ||
GenBank mRNA | AK033402.1 | - | 9 | 1277 | |
AK147064.1 | - | 9 | 1284 | ||
AK156055.1 | - | 9 | 1262 | ||
BC060124.1 | - | 9 | 1250 | 6408844 | |
BC069179.1 | - | 1 | 61 | ||
U27177.1 | - | 9 | 1267 | ||
U27178.1 | - | 9 | 1267 | ||
U33320.1 | - | 9 | 1250 |