Gene Transcripts

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
Chromosome Location
Chr. 3: 159988836 - 159996019 on Build GRCh38
UniGene ID
Hs.673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000882.3 NP_000873.2 6, 7 835
GenBank mRNA AF101062.1 - 6, 7 614
AF180562.1 - 6, 7 673
BC104982.1 - 6, 7 767 8143985
BC104984.1 - 6, 7 767 8143987
M65271.1 - 6, 7 681
M65291.1 - 6, 7 715

View Full Product Details