Gene Transcripts

Gene Symbol
St6galnac4
Entrez Gene ID
20448
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
Gene Aliases
SIAT7-D, ST6GalNAcIV, Siat7d
Chromosome Location
Chr. 2: 32587078 - 32599696 on Build GRCm38
UniGene ID
Mm.27446
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276425.1 NP_001263354.1 6 1022
NM_011373.3 NP_035503.1 5 1375
GenBank mRNA AJ007310.1 - 6 1017
AK007601.1 - 6 1004
AK085730.1 - 6 1013
AK144884.1 - 6 1013
AK153036.1 - 6 982
AK155956.1 - 4 3516
AK159712.1 - 6 1366
AK161559.1 - 6 1007
BC056451.1 - 5 828 5685866
BC086784.1 - 963 6844232
Y15779.1 - 6 999
Y15780.1 - 5 1011

View Full Product Details