Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079690.1 | NP_001073158.1 | 8, 9 | 1315 | |
NM_183354.2 | NP_899197.2 | 8, 9 | 1315 | ||
GenBank mRNA | BC016888.1 | - | 7, 8 | 1293 | |
BC051100.1 | - | 1, 2 | 79 | ||
U20973.1 | - | 8, 9 | 1313 | ||
U20974.1 | - | 8, 9 | 1313 | ||
U20975.1 | - | 8, 9 | 1313 | ||
U61381.1 | - | 7, 8 | 1284 | ||
U94518.1 | - | 7, 8 | 1262 |